MilliporeSigma
  • Thieno[2,3-d]pyrimidine-2-carboxamides bearing a carboxybenzene group at 5-position: highly potent, selective, and orally available MMP-13 inhibitors interacting with the S1″ binding site.
Thieno[2,3-d]pyrimidine-2-carboxamides bearing a carboxybenzene group at 5-position: highly potent, selective, and orally available MMP-13 inhibitors interacting with the S1″ binding site.
Bioorganic & medicinal chemistry (2014-09-07)
Hiroshi Nara, Kenjiro Sato, Takako Naito, Hideyuki Mototani, Hideyuki Oki, Yoshio Yamamoto, Haruhiko Kuno, Takashi Santou, Naoyuki Kanzaki, Jun Terauchi, Osamu Uchikawa, Masakuni Kori
ABSTRACT
On the basis of X-ray co-crystal structures of matrix metalloproteinase-13 (MMP-13) in complex with its inhibitors, our structure-based drug design (SBDD) strategy was directed to achieving high affinity through optimal protein-ligand interaction with the unique S1″ hydrophobic specificity pocket. This report details the optimization of lead compound 44 to highly potent and selective MMP-13 inhibitors based on fused pyrimidine scaffolds represented by the thienopyrimidin-4-one 26c. Furthermore, we have examined the release of collagen fragments from bovine nasal cartilage in response to a combination of IL-1 and oncostatin M.
Sigma-Aldrich
T-26c, ≥98% (HPLC)