MilliporeSigma
  • N-end rule pathway inhibition assists colon tumor regression via necroptosis.
N-end rule pathway inhibition assists colon tumor regression via necroptosis.
Molecular therapy oncolytics (2016-08-25)
Pritha Agarwalla, Rajkumar Banerjee
ABSTRACT
Recent study has shown that N-end rule pathway, an ubiquitin dependent proteolytic system, counteracts cell death by degrading many antisurvival protein fragments like BCLxL, BRCA1, RIPK1, etc. Inhibition of the N-end rule pathway can lead to metabolic stabilization of proapoptotic protein fragments like RIPK1, thereby sensitizing cells to programmed cell death. Receptor interacting serine-threonine protein kinase-1 (RIPK1) is one of the upstream regulators of programmed necrosis known as necroptosis. Necroptosis is particularly gaining attention of cancer biologists as it provides an alternate therapeutic modality to kill cancer cells, which often evolve multiple strategies to circumvent growth inhibition by apoptosis. Utilizing the over expression of biotin receptor in cancer cells, herein, we report that coadministration of synthetic hetero-bivalent N-end rule inhibitor RFC11 and anticancer drug shikonin solubilized in a stable biotin receptor-targeted liposome exhibited significant synergistic antitumor effect in both subcutaneous and orthotopic mouse colon tumor model through induction of necroptosis with distinctive upregulation of RIPK1. Besides developing a newly targeted formulation for necroptosis induction, this report is the first in vivo evidence demonstrating that potent inhibition of N-end rule pathway can enhance therapeutic efficacy of conventional chemotherapeutics.
Sigma-Aldrich
Goat Anti-Mouse IgG Alkaline Phosphatase Conjugate, lyophilized, Calbiochem®
Sigma-Aldrich
HBTU, ≥98.0% (T)