MilliporeSigma
  • Does Unc-GFP uncover ciliary structures in the rhabdomeric eye of Drosophila?
Does Unc-GFP uncover ciliary structures in the rhabdomeric eye of Drosophila?
Journal of cell science (2016-05-29)
Marco Gottardo, Giuliano Callaini, Maria Giovanna Riparbelli
ABSTRACT
The uncoordinated (unc) gene product, a potential ortholog of mammalian orofaciodigital syndrome 1 (Ofd1), is involved in the assembly of the ciliary axoneme in Drosophila and it is, therefore, constrained to cell types that have ciliary structures, namely type 1 sensory neurons and male germ cells. Here, we show that evenly spaced Unc-GFP spots are present in the eye imaginal discs of third-instar larvae. These spots are restricted to the R8 photoreceptor cell of each ommatidium in association with mother centrioles. This finding is unexpected because the Drosophila eye is of the rhabdomeric type and would be expected to lack ciliary structures.
Sigma-Aldrich
Monoclonal Anti-Tubulin, Acetylated antibody produced in mouse, clone 6-11B-1, ascites fluid
Sigma-Aldrich
Anti-CNTROB antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution, Ab1