MilliporeSigma
  • Targeted selected reaction monitoring mass spectrometric immunoassay for insulin-like growth factor 1.
Targeted selected reaction monitoring mass spectrometric immunoassay for insulin-like growth factor 1.
PloS one (2013-11-28)
Eric E Niederkofler, David A Phillips, Bryan Krastins, Vathany Kulasingam, Urban A Kiernan, Kemmons A Tubbs, Scott M Peterman, Amol Prakash, Eleftherios P Diamandis, Mary F Lopez, Dobrin Nedelkov
ABSTRACT
Insulin-like growth factor 1 (IGF1) is an important biomarker of human growth disorders that is routinely analyzed in clinical laboratories. Mass spectrometry-based workflows offer a viable alternative to standard IGF1 immunoassays, which utilize various pre-analytical preparation strategies. In this work we developed an assay that incorporates a novel sample preparation method for dissociating IGF1 from its binding proteins. The workflow also includes an immunoaffinity step using antibody-derivatized pipette tips, followed by elution, trypsin digestion, and LC-MS/MS separation and detection of the signature peptides in a selected reaction monitoring (SRM) mode. The resulting quantitative mass spectrometric immunoassay (MSIA) exhibited good linearity in the range of 1 to 1,500 ng/mL IGF1, intra- and inter-assay precision with CVs of less than 10%, and lowest limits of detection of 1 ng/mL. The linearity and recovery characteristics of the assay were also established, and the new method compared to a commercially available immunoassay using a large cohort of human serum samples. The IGF1 SRM MSIA is well suited for use in clinical laboratories.
Sigma-Aldrich
Albumin, Bovine Serum, Fraction V, Fatty Acid-Free, Nuclease- and Protease-Free, BSA Fatty Acid-free is designed for use in serological testing, RIA, and hormone response studies. Suitable for use in Molecular Biology applications, such as Northern and Southern blots.
Pricing and availability is not currently available.