MilliporeSigma
  • Urea uptake enhances barrier function and antimicrobial defense in humans by regulating epidermal gene expression.
Urea uptake enhances barrier function and antimicrobial defense in humans by regulating epidermal gene expression.
The Journal of investigative dermatology (2012-03-16)
Susanne Grether-Beck, Ingo Felsner, Heidi Brenden, Zippora Kohne, Marc Majora, Alessandra Marini, Thomas Jaenicke, Marina Rodriguez-Martin, Carles Trullas, Melanie Hupe, Peter M Elias, Jean Krutmann
ABSTRACT
Urea is an endogenous metabolite, known to enhance stratum corneum hydration. Yet, topical urea anecdotally also improves permeability barrier function, and it appears to exhibit antimicrobial activity. Hence, we hypothesized that urea is not merely a passive metabolite, but a small-molecule regulator of epidermal structure and function. In 21 human volunteers, topical urea improved barrier function in parallel with enhanced antimicrobial peptide (AMP; LL-37 and β-defensin-2) expression. Urea stimulates the expression of, and is transported into, keratinocytes by two urea transporters (UTs), UT-A1 and UT-A2, and by aquaporins 3, 7, and 9. Inhibitors of these UTs block the downstream biological effects of urea, which include increased mRNA and protein levels of (i) transglutaminase-1, involucrin, loricrin, and filaggrin, (ii) epidermal lipid synthetic enzymes, and (iii) cathelicidin/LL-37 and β-defensin-2. Finally, we explored the potential clinical utility of urea, showing that topical urea applications normalized both barrier function and AMP expression in a murine model of atopic dermatitis. Together, these results show that urea is a small-molecule regulator of epidermal permeability barrier function and AMP expression after transporter uptake, followed by gene regulatory activity in normal epidermis, with potential therapeutic applications in diseased skin.
Sigma-Aldrich
MISSION® esiRNA, targeting human FLG
Sigma-Aldrich
MISSION® esiRNA, targeting human IVL