MilliporeSigma
  • A thermostable β-glucuronidase obtained by directed evolution as a reporter gene in transgenic plants.
A thermostable β-glucuronidase obtained by directed evolution as a reporter gene in transgenic plants.
PloS one (2011-11-19)
Ai-Sheng Xiong, Ri-He Peng, Jing Zhuang, Jian-Min Chen, Bin Zhang, Jian Zhang, Quan-Hong Yao
ABSTRACT
A β-glucuronidase variant, GUS-TR3337, that was obtained by directed evolution exhibited higher thermostability than the wild-type enzyme, GUS-WT. In this study, the utility of GUS-TR337 as an improved reporter was evaluated. The corresponding gus-tr3337 and gus-wt genes were independently cloned in a plant expression vector and introduced into Arabidopsis thaliana. With 4-MUG as a substrate, plants containing the gus-wt gene showed no detectable β-glucuronidase activity after exposure to 60°C for 10 min, while those hosting the gus-tr3337 gene retained 70% or 50% activity after exposure to 80°C for 10 min or 30 min, respectively. Similarly, in vivo β-glucuronidase activity could be demonstrated by using X-GLUC as a substrate in transgenic Arabidopsis plants hosting the gus-tr3337 gene that were exposed to 80°C for up to 30 min. Thus, the thermostability of GUS-TR3337 can be exploited to distinguish between endogenous and transgenic β-glucuronidase activity, which is a welcome improvement in its use as a reporter.
Sigma-Aldrich
4-Methylumbelliferone sodium salt, ≥98% (HPLC), crystalline