MilliporeSigma
  • Cycloadditions of 1,2,3-Triazines Bearing C5-Electron Donating Substituents: Robust Pyrimidine Synthesis.
Cycloadditions of 1,2,3-Triazines Bearing C5-Electron Donating Substituents: Robust Pyrimidine Synthesis.
Organic letters (2015-07-15)
Christopher M Glinkerman, Dale L Boger
ABSTRACT
The examination of the cycloaddition reactions of 1,2,3-triazines 17-19, bearing electron-donating substituents at C5, are described. Despite the noncomplementary 1,2,3-triazine C5 substituents, amidines were found to undergo a powerful cycloaddition to provide 2,5-disubstituted pyrimidines in excellent yields (42-99%; EDG = SMe > OMe > NHAc). Even select ynamines and enamines were capable of cycloadditions with 17, but not 18 or 19, to provide trisubstituted pyridines in modest yields (37-40% and 33% respectively).
Sigma-Aldrich
5-Methoxy-1,2,3-triazine