MilliporeSigma
  • Bespoke SnAP reagents for the synthesis of C-substituted spirocyclic and bicyclic saturated N-heterocycles.
Bespoke SnAP reagents for the synthesis of C-substituted spirocyclic and bicyclic saturated N-heterocycles.
Organic letters (2015-03-31)
Kimberly Geoghegan, Jeffrey W Bode
ABSTRACT
The precise placement of C-substituents on bicyclic and spirocyclic N-heterocycles is readily achieved by the combination of aldehydes and new SnAP reagents. The substituted SnAP reagents are readily prepared from simple starting materials and couple with a variety of aromatic and heteroaromatic aldehydes at room temperature under operationally simple conditions to deliver substituted morpholine and piperazine products.