MilliporeSigma
  • Novel process for producing 6-deoxy monosaccharides from l-fucose by coupling and sequential enzymatic method.
Novel process for producing 6-deoxy monosaccharides from l-fucose by coupling and sequential enzymatic method.
Journal of bioscience and bioengineering (2015-06-03)
Sirinan Shompoosang, Akihide Yoshihara, Keiko Uechi, Yasuhiko Asada, Kenji Morimoto
ABSTRACT
We biosynthesized 6-deoxy-L-talose, 6-deoxy-L-sorbose, 6-deoxy-L-gulose, and 6-deoxy-L-idose, which rarely exist in nature, from L-fucose by coupling and sequential enzymatic reactions. The first product, 6-deoxy-L-talose, was directly produced from L-fucose by the coupling reactions of immobilized D-arabinose isomerase and immobilized L-rhamnose isomerase. In one-pot reactions, the equilibrium ratio of L-fucose, L-fuculose, and 6-deoxy-L-talose was 80:9:11. In contrast, 6-deoxy-L-sorbose, 6-deoxy-L-gulose, and 6-deoxy-L-idose were produced from L-fucose by sequential enzymatic reactions. D-Arabinose isomerase converted L-fucose into L-fuculose with a ratio of 88:12. Purified L-fuculose was further epimerized into 6-deoxy-L-sorbose by D-allulose 3-epimerase with a ratio of 40:60. Finally, purified 6-deoxy-L-sorbose was isomerized into both 6-deoxy-L-gulose with an equilibrium ratio of 40:60 by L-ribose isomerase, and 6-deoxy-L-idose with an equilibrium ratio of 73:27 by D-glucose isomerase. Based on the amount of L-fucose used, the production yields of 6-deoxy-L-talose, 6-deoxy-L-sorbose, 6-deoxy-L-gulose, and 6-deoxy-L-idose were 7.1%, 14%, 2%, and 2.4%, respectively.
Sigma-Aldrich
L-(+)-Ribose, ≥98% (GC)
Sigma-Aldrich
L-Rhamnose, natural sourced, 99%, FG