MilliporeSigma
  • Andrographolide suppress tumor growth by inhibiting TLR4/NF-κB signaling activation in insulinoma.
Andrographolide suppress tumor growth by inhibiting TLR4/NF-κB signaling activation in insulinoma.
International journal of biological sciences (2014-04-11)
Qian-Qian Zhang, Yi Ding, Yan Lei, Cui-Ling Qi, Xiao-Dong He, Tian Lan, Jiang-Chao Li, Ping Gong, Xuesong Yang, Jian-Guo Geng, Li-Jing Wang
ABSTRACT
Insulinomas are rare tumors, and approximately 10% of insulinomas are malignant. Accumulating evidence has implicated that we still lack effective therapy to treat the patients who are diagnosed with rare malignant insulinoma. Previous studies have reported that Andrographolide (Andro) could inhibit cell cycle progression, reduce cell invasion and induce cell apoptosis in many common cancer cells. However, the effects of andro are cell type-dependent. So we emplored the β-TC-6 cells and the RIP1-Tag2 transgenic mouse model of endogenously growing insulinoma model to elucidate the possible anti-cancer effect of Andro on insulinoma, an uncommon type of malignant cancers in this study. Our experiments revealed that Andro significantly inhibited tumor growth at both the early-stage and the advanced-stage of insulinoma through targeting the TLR4/NF-κB signaling pathway. This work initially provides the evidence that the TLR4/NF-κB signaling pathway might be vital as a potential therapeutic target, and also indispensable in Andro-mediated anti-cancer effect in insulinoma.
Product Description
Roche
In Situ Cell Death Detection Kit, AP, sufficient for ≤50 tests, kit of 1 (3 components), suitable for detection