MilliporeSigma
Effects of Antitumor Drug Sorafenib on Chick Embryo Development.
Anatomical record (Hoboken, N.J. : 2007) (2015-03-27)
Yi-Sen Cheng, Xiao-Yu Wang, Guang Wang, Yan Li, Yue-Lei Chen, Man-Li Chuai, Kenneth Ka Ho Lee, Xiao-Yan Ding, Xue-Song Yang
ABSTRACT
Sorafenib has been used as an oral anti-cancer drug because of its ability to inhibit tumor growth. However, the pharmacological effect of sorafenib is still the lack of in vivo experimental evidence. Tumor and embryonic cells share some similar features, so we investigated the effects of sorafenib on the development of gastrulating chick embryos. We found that sorafenib exposure was markedly attributed to the number of embryonic cell in proliferation and apoptosis. We also detected sorafenib significantly interfered with epithelial-mesenchymal transition (EMT). Furthermore, sorafenib treatment impaired the production and migration of neural crest cells.
Sigma-Aldrich
Monoclonal Anti-HNK-1/N-CAM (CD57) antibody produced in mouse, clone VC1.1, ascites fluid, buffered aqueous solution