MilliporeSigma
  • Click chemistry modification of natural keratin fibers for sustained shrink-resist performance.
Click chemistry modification of natural keratin fibers for sustained shrink-resist performance.
International journal of biological macromolecules (2015-04-04)
Dan Yu, Jackie Y Cai, Jeffrey S Church, Lijing Wang
ABSTRACT
This paper introduces a novel chemical treatment for achieving sustained shrink-resist performance on natural keratin fibers. The new treatment involves the controlled reduction of keratin in the cuticle region of the fiber, and the application of a water soluble diacrylate, namely glycerol 1,3-diglycerolate diacrylate (GDA), on the reduced keratin substrate. The acrylate groups of the GDA react with cysteine residues in the reduced keratin through thiol-ene click reactions at room temperature, leading to GDA grafting and the formation of GDA crosslinks in the keratin structure. The modified substrates were characterized by infrared spectroscopy and scanning electron microscopy, and assessed for its shrink-resistance and wet burst strength. This chemical modification has shown to alter the fiber surface morphology and hydrophilicity, resulting in substantially improved shrink-resistance with good fiber strength retention. Possible shrink-resistance mechanisms were also discussed.
Sigma-Aldrich
Glycerol 1,3-diglycerolate diacrylate, technical grade
Sigma-Aldrich
Nitrogen, ≥99.998%