MilliporeSigma
  • Functional role of the RET dependence receptor, GFRa co-receptors and ligands in the pituitary.
Functional role of the RET dependence receptor, GFRa co-receptors and ligands in the pituitary.
Frontiers of hormone research (2010-07-10)
Montserrat Garcia-Lavandeira, Esther Diaz-Rodriguez, Maria E R Garcia-Rendueles, Joana S Rodrigues, Sihara Perez-Romero, Susana B Bravo, Clara V Alvarez
ABSTRACT
The RET receptor is a tyrosine kinase receptor implicated in kidney and neural development. In the adenopituitary RET and the co-receptor GFRa1 are expressed exclusively in the somatotrophs secreting GH. RET is implicated in a clever pathway to maintain at physiological levels the number of somatotrophs and the GH production. Thus, in absence of its ligand GDNF, RET induces apoptosis through massive expression of Pit-1 leading to p53 accumulation. In the presence of the ligand GDNF, RET activates its tyrosine kinase and promotes survival at the expense of reducing Pit-1 expression and downregulating GH. Recent data suggest that RET can also have a second role in pituitary plasticity through a second co-receptor GFRa2.