MilliporeSigma
  • Oscillometric casual blood pressure normative standards for Swedish children using ABPM to exclude casual hypertension.
Oscillometric casual blood pressure normative standards for Swedish children using ABPM to exclude casual hypertension.
American journal of hypertension (2014-11-12)
Rafael T Krmar, Ulla Holtbäck, Anita Bergh, Eva Svensson, Elke Wühl
ABSTRACT
Casual blood pressure (CBP) is considered a reliable proxy for cardiovascular health. Although the auscultatory technique is the reference standard method for measuring CBP, oscillometric devices are increasingly being used in children. We sought to establish oscillometric CBP normative standards for Swedish children. Cross-sectional oscillometric CBP readings were obtained by the Welch Allyn Spot Vital Signs 420 monitor and measured according to the International Guidelines' recommendations. Participants with elevated oscillometric CBP levels underwent verification by the auscultatory method. Ambulatory blood pressure monitoring (ABPM) was used to exclude casual hypertension. Data on 1,470 (772 males) apparently healthy Swedish schoolchildren aged 6-16 years were analyzed and sex-specific reference charts normalized to age or height were constructed. Systolic and diastolic CBP values were significantly higher with age, height, height standard deviation score (SDS), body mass index (BMI), and BMI SDS. Gender differences for systolic CBP were present starting from age of 15 years and revealed significantly higher values in boys than in girls, whereas for diastolic CBP, the differences were apparent at the age of 12 years, with higher values in girls. Increased BMI and BMI SDS were positively associated with CBP levels. Positive parental history of hypertension turned out to be a risk factor for higher systolic and diastolic CBP across all ages. Our normative standard for CBP can be used for blood pressure screening and control programs in Swedish children. The use of ABPM should be considered to confirm the diagnosis of casual hypertension.
Sigma-Aldrich
4,4′-Bis(N-carbazolyl)-1,1′-biphenyl, 97%
Sigma-Aldrich
4,4′-Bis(N-carbazolyl)-1,1′-biphenyl, sublimed grade, 99.9% trace metals basis