MilliporeSigma
  • Water ice is a soft matrix for the structural characterization of glycosaminoglycans by infrared matrix-assisted laser desorption/ionization.
Water ice is a soft matrix for the structural characterization of glycosaminoglycans by infrared matrix-assisted laser desorption/ionization.
Analytical chemistry (2014-05-28)
Lukas Witt, Alexander Pirkl, Felix Draude, Jasna Peter-Katalinić, Klaus Dreisewerd, Michael Mormann
ABSTRACT
Glycosaminoglycans (GAGs) are a class of heterogeneous, often highly sulfated glycans that form linear chains consisting of up to 100 monosaccharide building blocks and more. GAGs are ubiquitous constituents of connective tissue, cartilage, and the extracellular matrix, where they have key functions in many important biological processes. For their characterization by mass spectrometry (MS) and tandem MS, the high molecular weight polymers are usually enzymatically digested to oligomers with a low degree of polymerization (dp), typically disaccharides. However, owing to their lability elimination of sulfate groups upon desorption/ionization is often encountered leading to a loss of information on the analyte. Here, we demonstrate that, in particular, water ice constitutes an extremely mild matrix for the analysis of highly sulfated GAG disaccharides by infrared matrix-assisted laser desorption/ionization (IR-MALDI) mass spectrometry. Depending on the degree of sulfation, next to the singly charged ionic species doubly- and even triply charged ions are formed. An unambiguous assignment of the sulfation sites becomes possible by subjecting sodium adducts of the GAGs to low-energy collision-induced dissociation tandem MS. These ionic species exhibit a remarkable stability of the sulfate substituents, allowing the formation of fragment ions retaining their sulfation that arise from either cross-ring cleavages or rupture of the glycosidic bonds, thereby allowing an unambiguous assignment of the sulfation sites.
Product Description
Supelco
Calcium Acetate, Pharmaceutical Secondary Standard; Certified Reference Material