MilliporeSigma
  • [Association of the glycophorin A gene mutation in peripheral erythrocytes and chronic benzene poisoning].
[Association of the glycophorin A gene mutation in peripheral erythrocytes and chronic benzene poisoning].
Zhonghua lao dong wei sheng zhi ye bing za zhi = Zhonghua laodong weisheng zhiyebing zazhi = Chinese journal of industrial hygiene and occupational diseases (2007-04-26)
Cai-hong Xing, Zhi-ying Ji, Gui-lan Li, Song-nian Yin
ABSTRACT
To explore the association of the glycophorin A(GPA) gene mutation in peripheral erythrocytes and chronic benzene poisoning. Sixty-three patients with chronic benzene poisonings and 45 benzene-exposed workers who were engaged in the same job title were investigated. Fluorescence immunolabeling technique and flow cytometry were used to detect GPA mutation frequency in peripheral read blood cell. A significant decrease in WBC count and neutrophil count was found in patients with chronic benzene poisoning compared with control individuals (P<0.01). The WBC count and neutrophil count both decreased along with the GPA-NN frequency, and the trends were significant(P<0.05).Both WBC counts and neutrophil counts decreased as the frequency, and trends were significant(P<0.05). GPA-NN frequency increased along with the accumulative exposure score, and the trend was significant (P = 0.0026). There was no significant trend between the GPA-Nphi frequency and the accumulative exposure score (P = 0.2037). A decrease in WBC count and neutrophil count is found in patients with chronic benzene poisoning, which can arise from genetic damage in bone marrow stem cells, namely gene-duplicating mutations (NN) at the GPA locus in bone marrow cells of MN-heterozygous subjects, GPA-NN mutagens contributed to the pathogenesis of chronic benzene poisoning.