MilliporeSigma
  • The molecular mechanism for human metallothionein-3 to protect against the neuronal cytotoxicity of Aβ(1-42) with Cu ions.
The molecular mechanism for human metallothionein-3 to protect against the neuronal cytotoxicity of Aβ(1-42) with Cu ions.
Journal of biological inorganic chemistry : JBIC : a publication of the Society of Biological Inorganic Chemistry (2012-10-23)
Ying Luo, Yuxia Xu, Qingui Bao, Zhichun Ding, Cuiqing Zhu, Zhong-Xian Huang, Xiangshi Tan
ABSTRACT
Aggregation and cytotoxicity of Aβ with redox-active metals in neuronal cells have been implicated in the progression of Alzheimer disease. Human metallothionein (MT) 3 is highly expressed in the normal human brain and is downregulated in Alzheimer disease. Zn(7)MT3 can protect against the neuronal toxicity of Aβ by preventing copper-mediated Aβ aggregation, abolishing the production of reactive oxygen species (ROS) and the related cellular toxicity. In this study, we intended to decipher the roles of single-domain proteins (α/β) and the α-β domain-domain interaction of Zn(7)MT3 to determine the molecular mechanism for protection against the neuronal cytotoxicity of Aβ(1-42) with copper ions. With this in mind, the α and β single-domain proteins, heterozygous β(MT3)-α(MT1), and a linker-truncated mutant ∆31-34 were prepared and characterized. In the presence/absence of various Zn(7)MT3 proteins, the Aβ(1-42)-Cu(2+)-mediated aggregation, the production of ROS, and the cellular toxicity were investigated by transmission electron microscopy, ROS assay by means of a fluorescent probe, and SH-SY5Y cell viability, respectively. The β domain cannot abolish Aβ(1-42)-Cu(2+)-induced aggregation, and neither the β domain nor the α domain can quench the production of ROS because of the redox cycling of Aβ-Cu(2+). Similarly to wild-type Zn(7)MT3, the heterozygous β(MT3)-α(MT1) possesses the characteristic of alleviating Aβ(1-42) aggregation and oxidative stress to neuronal cells. Therefore, the two domains through the linker Lys-Lys-Ser form a cooperative unit, and each of them is indispensable in conducting its bioactivity. The α domain plays an important role in modulating the stability of the metal-thiolate cluster, and the α-β domain-domain interaction through the linker is critical for its protective role in the brain.