MilliporeSigma
  • Netupitant and palonosetron trigger NK1 receptor internalization in NG108-15 cells.
Netupitant and palonosetron trigger NK1 receptor internalization in NG108-15 cells.
Experimental brain research (2014-06-28)
Ajit G Thomas, Marigo Stathis, Camilo Rojas, Barbara S Slusher
ABSTRACT
Current therapy for chemotherapy-induced nausea and vomiting includes the use of both 5-HT3 and NK1 receptor antagonists. Acute emesis has largely been alleviated with the use of 5-HT3 receptor antagonists, while an improvement in preventing delayed emesis has been achieved with NK1 receptor antagonists. Delayed emesis, however, remains a problem with a significant portion of cancer patients receiving highly emetogenic chemotherapy. Like other drugs in its class, palonosetron, a 5-HT3 receptor antagonist, has shown efficacy against acute emesis. However, palonosetron has also shown consistent improvement in the suppression of delayed emesis. Since both 5-HT3 and NK1 receptor antagonists are often simultaneously administered to patients, the question remains if palonosetron's effect on delayed emesis would remain distinct when co-administered with an NK1 receptor antagonist. Recent mechanistic studies using NG108-15 cells have shown that palonosetron and netupitant, an NK1 receptor antagonist currently in phase 3 clinical trials, exhibited synergistic effects when inhibiting the substance P response. The present studies showed that both netupitant and palonosetron-induced NK1 receptor internalization in NG108-15 cells and that when used together receptor internalization was additive. Palonosetron-induced NK1 receptor internalization was dependent on the presence of the 5-HT3 receptor. Results provide a possible explanation for palonosetron's enhancement of the inhibition of the SP response and suggest that the effect of palonosetron and NK1 receptor antagonists on prevention of delayed emesis could be additive.
Sigma-Aldrich
Netupitant, ≥98% (HPLC)