MilliporeSigma
  • CD34 and factor XIIIa in the differential diagnosis of dermatofibroma and dermatofibrosarcoma protuberans.
CD34 and factor XIIIa in the differential diagnosis of dermatofibroma and dermatofibrosarcoma protuberans.
The American Journal of dermatopathology (1993-10-01)
P Abenoza, T Lillemoe
ABSTRACT
To assess the use of anti-CD34 and anti-factor-XIIIa antibodies for the differential diagnosis of dermatofibroma (DF) and dermatofibrosarcoma protuberans (DFSP), we stained 40 DFs and 13 DFSPs. A significant population of dendritic and spindle cells was reactive with anti-factor-XIIIa in 90% of DFs. In DFSP, most cases had very few or no reactive cells for this antiserum and reactivity was confined to the infiltrating edges of the neoplasm or to a very low percentage of cells within the bulk of the tumor. A significant overlap was detected, a reason why the stain is useful but lacks absolute specificity for this purpose. Human progenitor cell antigen (CD34) was found in all cases of DFSP that had 50-100% of reactive cells, but in only five DFs. In DFs, the reactivity for CD34 was focal, with only < or = 20% positive cells. When used together with other histopathological criteria, factor-XIIIa antibody and particularly CD34 may help to differentiate these two processes.
Sigma-Aldrich
Factor XIIIa (AC-1A1) Mouse Monoclonal Antibody
Sigma-Aldrich
Factor XIIIa (EP3372) Rabbit Monoclonal Primary Antibody