MilliporeSigma
  • Myoepithelial tumors of soft tissue: a clinicopathologic and immunohistochemical study of 101 cases with evaluation of prognostic parameters.
Myoepithelial tumors of soft tissue: a clinicopathologic and immunohistochemical study of 101 cases with evaluation of prognostic parameters.
The American journal of surgical pathology (2003-09-10)
Jason L Hornick, Christopher D M Fletcher
ABSTRACT
Myoepitheliomas and mixed tumors were only recently recognized to occur primarily in soft tissue, and only small case numbers have been described. To characterize these tumors further and to evaluate prognostic parameters, 101 myoepithelial tumors of soft tissue were retrieved from the authors' consult files. Hematoxylin and eosin sections were reexamined, immunohistochemistry was performed, and clinical details were obtained from referring physicians. Fifty-three patients were male and 48 female (mean age 38 years; range 3-83 years). Tumor size ranged from 0.7 to 20 cm (mean 4.7 cm). Most tumors arose in the extremities and limb girdles: 41 in the lower limbs, 35 in the upper limbs, 15 in the head and neck, and 10 in the trunk. Fifty-four tumors were situated in subcutis and 37 in deep soft tissue (depth unstated in 10). Most cases were grossly well circumscribed; 43 showed microscopically infiltrative margins. Histologically, most tumors were lobulated, composed of cords or nests of epithelioid, ovoid, or spindled cells with a variably reticular architecture and a chondromyxoid or collagenous/hyalinized stroma. Eight cases showed a predominantly solid proliferation of spindled or plasmacytoid cells; 17 demonstrated ductular differentiation (mixed tumors). Cartilage was present in 6 cases, 6 contained bone, and 4 others contained both. Mitoses ranged from 0 to 68 per 10 high power fields (mean 4.7 per 10 high power fields). Tumors with benign cytomorphology or mild cytologic atypia (low-grade) were classified as myoepithelioma or mixed tumor, whereas tumors with moderate to severe atypia (high-grade) were classified as myoepithelial carcinoma (epithelioid or spindled cells with vesicular or coarse chromatin, prominent, often large nucleoli, or nuclear pleomorphism) or malignant mixed tumor (cytologically malignant cartilage or bone). Sixty-one cases were myoepitheliomas or mixed tumors, and 40 were myoepithelial carcinomas or malignant mixed tumors. By immunohistochemistry, all cases with available material were reactive for epithelial markers (keratins and/or epithelial membrane antigen): 90 of 97 (93%) expressed keratins (most often AE1/AE3 or PAN-K), 84 of 97 (87%) S-100 protein, 44 of 51 (86%) calponin, 52 of 83 (63%) epithelial membrane antigen, 40 of 87 (46%) glial fibrillary acidic protein, 27 of 75 (36%) smooth muscle actin, 15 of 66 (23%) p63, and 7 of 51 (14%) desmin. Follow-up was available for 64 patients. Among 33 cases with benign or low-grade cytology (mean follow-up 36 months; range 4-168 months), 6 recurred locally (18%) and none metastasized. No clinical or histologic features correlated with recurrence. Among 31 cytologically malignant cases (mean follow-up 50 months; range 4-252 months), 13 recurred locally (42%) and 10 metastasized (32%); so far, 4 patients have died of metastatic tumor. This study expands the spectrum of myoepithelial tumors of soft tissue to include myoepithelial carcinomas and malignant mixed tumors, which pursue an aggressive clinical course. Although the majority of morphologically benign or low-grade myoepithelial neoplasms of soft tissue behave in a benign fashion, there is an approximate 20% risk for local recurrence.
Sigma-Aldrich
Calponin-1 (EP798Y) Rabbit Monoclonal Primary Antibody
Sigma-Aldrich
Calponin (CALP) Mouse Monoclonal Antibody