MilliporeSigma
  • Directed evolution of epoxide hydrolase from A. radiobacter toward higher enantioselectivity by error-prone PCR and DNA shuffling.
Directed evolution of epoxide hydrolase from A. radiobacter toward higher enantioselectivity by error-prone PCR and DNA shuffling.
Chemistry & biology (2004-07-24)
Bert van Loo, Jeffrey H Lutje Spelberg, Jaap Kingma, Theo Sonke, Marcel G Wubbolts, Dick B Janssen
ABSTRACT
The enantioselectivity of epoxide hydrolase from Agrobacterium radiobacter (EchA) was improved using error-prone PCR and DNA shuffling. An agar plate assay was used to screen the mutant libraries for activity. Screening for improved enantioselectivity was subsequently done by spectrophotometric progress curve analysis of the conversion of para-nitrophenyl glycidyl ether (pNPGE). Kinetic resolutions showed that eight mutants were obtained with up to 13-fold improved enantioselectivity toward pNPGE and at least three other epoxides. The large enhancements in enantioselectivity toward epichlorohydrin and 1,2-epoxyhexane indicated that pNPGE acts as an epoxyalkane mimic. Active site mutations were found in all shuffled mutants, which can be explained by an interaction of the affected amino acid with the epoxide oxygen or the hydrophobic moiety of the substrate. Several mutations in the shuffled mutants had additive effects.
Product Description
Supelco
Astec® CHIRALDEX G-TA Capillary GC Column, L × I.D. 30 m × 0.25 mm, df 0.12 μm