MilliporeSigma
Covalent bonding of azoles to quaternary protoberberine alkaloids.
Magnetic resonance in chemistry : MRC (2008-09-11)
Lenka Grycová, Dagmar Hulová, Lukás Maier, Stanislav Standara, Marek Necas, Filip Lemière, Radovan Kares, Jirí Dostál, Radek Marek
ABSTRACT
Adducts of the quaternary protoberberine alkaloids (QPA) berberine, palmatine, and coptisine were prepared with nucleophiles derived from pyrrole, pyrazole, imidazole, and 1,2,4-triazole. The products, 8-substituted 7,8-dihydroprotoberberines, were identified by mass spectrometry and 1D and 2D NMR spectroscopy, including (1)H--(15)N shift correlations at natural abundance. In addition, two adducts of QPA with chloroform and methanethiolate were characterized by using NMR data. Single-crystal X-ray structures of 8-pyrrolyl-7,8-dihydroberberine, 8-pyrazolyl-7,8-dihydroberberine, and 8-imidazolyl-7,8-dihydroberberine are also presented.
Sigma-Aldrich
Palmatine chloride hydrate, 97%