MilliporeSigma
Packing columns for capillary electrochromatography.
Journal of chromatography. A (2000-08-29)
L A Colón, T D Maloney, A M Fermier
ABSTRACT
Considering the current interest in capillary electrochromatography (CEC), performed in packed columns, we present the different methods used to pack capillary columns for use in CEC. General considerations on column packing are given and the column fabrication process is discussed in sufficient detail to allow instruction to those who are not experienced in the field. Five different packing methods are discussed to deliver packing material into the capillary column from a practical view point: slurry pressure packing, packing with supercritical CO2, electrokinetic packing, using centripetal forces, and packing by gravity. Entrapment of particulate material by sintering and sol-gel technology is also mentioned. Although slurry pressure packing procedures are most common, higher separation efficiencies are obtained using other packing approaches. Electrokinetic packing seems to be the simplest technique to deliver the packing material into the capillary columns. Nevertheless, as with the other packing techniques, skill and experience are required to complete all the steps involved in the fabrication of packed columns for CEC.
Product Description
Frit for Omnifit® Column Endpiece, PTFE, for use with 10 mm bore, pore size 10 μm
Frit for Omnifit® Column Endpiece, polyethylene, for use with 10 mm Bore Columns, pore size 25 μm