MilliporeSigma
  • The genetic toxicity effects of lamivudine and stavudine antiretroviral agents.
The genetic toxicity effects of lamivudine and stavudine antiretroviral agents.
Expert opinion on drug safety (2010-04-10)
Nilza Nascimento Guimarães, Heloísa Helena Rodrigues de Andrade, Maurício Lehmann, Rafael Rodrigues Dihl, Kênya Silva Cunha
ABSTRACT
The nucleoside reverse transcriptase inhibitors (NRTIs) are used in antiretroviral therapy worldwide for the treatment of HIV infections. These drugs act by blocking reverse transcriptase enzyme activity, causing pro-viral DNA chain termination. As a consequence, NRTIs could cause genomic instability and loss of heterozygosity. This review highlights the toxic and genotoxic effects of NRTIs, particularly lamivudine (3TC) and stavudine (d4T) analogues. In addition, a battery of short-term in vitro and in vivo systems are described to explain the potential genotoxic effects of these NRTIs as a single drug or a complexity of highly active antiretroviral therapy. The readers will gain an understanding of a secondary effect that could be induced by 3TC and d4T treatments. Considering that AIDS has become a chronic disease, more comprehensive toxic genetic studies are needed, with particular attention to the genetic alterations induced by NRTIs. These alterations play a primary role in carcinogenesis and are also involved in secondary and subsequent steps of carcinogenesis.
Product Description
Stavudine for system suitability, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
2′,3′-Didehydro-3′-deoxythymidine, ≥98% (TLC)
Stavudine, European Pharmacopoeia (EP) Reference Standard