MilliporeSigma
  • Hydrothermally mixed hydroxyapatite-multiwall carbon nanotubes composite coatings on biomedical alloys by electrophoretic deposition.
Hydrothermally mixed hydroxyapatite-multiwall carbon nanotubes composite coatings on biomedical alloys by electrophoretic deposition.
The journal of physical chemistry. B (2012-07-12)
C B Ustundag, O Avciata, F Kaya, C Kaya
ABSTRACT
Hydroxyapatite (HA) coatings have been used to improve biological and mechanical fixation of metallic prosthesis. Because of extraordinary features of carbon nanotubes (CNTs), they have a lot of facilities, such as extremely strong nanoreinforcement materials for composites. HA powders were synthesized and mixed with multiwalled carbon nanotubes (MWCNTs) by a hydrothermal process. Calcium acetate (Ca (CH(3)COO)(2)) and phosphoric acid (H(3)PO(4)) were used as starting materials for synthesizing nano-HA powders. HA-MWCNTs were treated together hydrothermally at 200 °C for 2 h to synthesize nano-HA powders mixed homogeneously with MWCNTs. Cathodic deposits were obtained on Ti-based alloys using suspensions containing nano-HA and MWCNTs dispersed in n-butanol solvent. It was shown that MWCNTs interacted with HA powders during hydrothermal processing, and therefore, they can easily be dispersed within aqueous-based suspensions. It was also shown that hydrothermal surface modification of MWCNTs with functional groups was achievable, which was a significant step toward eliminating nonwetting surface behavior of MWCNTs, resulting in obtaining homogeneous dispersion of them in liquids.
Product Description
Supelco
Calcium Acetate, Pharmaceutical Secondary Standard; Certified Reference Material