MilliporeSigma
  • Safety of long-term treatment with cabergoline on cardiac valve disease in patients with prolactinomas.
Safety of long-term treatment with cabergoline on cardiac valve disease in patients with prolactinomas.
European journal of endocrinology (2013-07-05)
Renata S Auriemma, Rosario Pivonello, Ylenia Perone, Ludovica F S Grasso, Lucia Ferreri, Chiara Simeoli, Davide Iacuaniello, Maurizio Gasperi, Annamaria Colao
ABSTRACT
Cabergoline (CAB) has been found to be associated with increased risk of cardiac valve regurgitation in Parkinson's disease, whereas several retrospective analyses failed to detect a similar relation in hyperprolactinemic patients. The current study aimed at investigating cardiac valve disease before and after 24 and 60 months of continuous treatment with CAB only in patients with hyperprolactinemia. Forty patients (11 men and 29 women, aged 38.7 ± 12.5 years) newly diagnosed with hyperprolactinemia entered the study. Cumulative CAB dose ranged from 12 to 588 mg (median 48 mg) at 24 months and 48-1260 mg (median 149 mg) at 60 months. All patients underwent a complete trans-thoracic echocardiographic examination. Valve regurgitation was assessed according to the American Society of Echocardiography. At baseline, the prevalence of trace mitral, aortic, pulmonic, and tricuspid regurgitations was 20, 2.5, 10, and 40% respectively, with no patient showing clinically relevant valvulopathy. After 24 months, no change in the prevalence of trace mitral (P=0.78) and pulmonic (P=0.89) regurgitations and of mild aortic (P=0.89) and tricuspid (P=0.89) regurgitations was found when compared with baseline. After 60 months, the prevalence of trace tricuspid regurgitation was only slightly increased when compared with that after 24 months (37.5%; P=0.82), but none of the patients developed significant valvulopathy. No correlation was found between cumulative dose and prevalence or grade of valve regurgitation at both evaluations. Prolactin levels normalized in all patients but one. CAB does not increase the risk of significant cardiac valve regurgitation in prolactinomas after the first 5 years of treatment.
Sigma-Aldrich
Cabergoline, ≥98% (HPLC)
Cabergoline, European Pharmacopoeia (EP) Reference Standard