MilliporeSigma
D-glycerate kinase deficiency as a cause of D-glyceric aciduria.
FEBS letters (1989-01-30)
E Van Schaftingen
ABSTRACT
D-Glycerate kinase was measured in human livers thanks to a new, sensitive radiochemical assay. The enzyme was extremely unstable in extracts prepared in water, but was partly stabilized in a homogenization mixture containing inorganic phosphate, D-glycerate and EGTA. When extracted in such a stabilizing mixture, glycerate kinase activity amounted to 0.86 +/- 0.21 U/g in control livers and to 0.03 U/g in the liver of a patient with D-glyceric aciduria. In contrast, D-glycerate dehydrogenase (glyoxylate reductase) and triokinase activities were not deficient in the liver of the same patient. It is concluded that D-glycerate kinase deficiency is a cause of D-glyceric aciduria.
Sigma-Aldrich
D-Glyceric acid sodium salt, ≥95.0% (TLC)