MilliporeSigma
  • Effects of chronic oral treatment with GABA-transaminase inhibitors on the GABA system in brain, liver, kidney, and plasma of the rat.
Effects of chronic oral treatment with GABA-transaminase inhibitors on the GABA system in brain, liver, kidney, and plasma of the rat.
Biochemical pharmacology (1996-11-08)
M Qume, L J Fowler
ABSTRACT
The inhibitory neurotransmitter gamma-aminobutyric acid (GABA) is not solely located in the CNS, it and the enzymes responsible for its synthesis (glutamic acid decarboxylase, GAD, EC 4.1.1.15) and catabolism (GABA-transaminase, GABA-T, EC 2.6.1.19) are also present in non-neuronal organs. Following 2, 8 and 21 day oral administration of ethanolamine-O-sulphate (EOS) and gamma-vinyl GABA (GVG), two irreversible inhibitors of GABA-T, the GABA content and activities of GAD and GABA-T in rat brain, liver and kidney, and the GABA content of plasma were determined: GABA-T activity was significantly decreased (over 80%) in liver, brain and kidney, although there was 2-3 times the residual activity left in the brain compared with the peripheral organs. GABA content was subsequently significantly elevated in the liver (300-1500%), plasma (200-300%) and brain (200-300%), although, surprisingly, the kidney GABA content was reduced (by 60-70%) compared with control. GAD activity was decreased following 8 day treatment in liver and brain. Kidney GAD was reduced at all time points. These two compounds are anticonvulsant, GVG is used clinically for the treatment of epilepsy but it seems that these drugs have significant peripheral effects.
Sigma-Aldrich
2-Aminoethyl hydrogen sulfate, ≥98.0% (T)