MilliporeSigma
  • Residues of cycloate and influence of herbicides on some of nutritional factors in spinach.
Residues of cycloate and influence of herbicides on some of nutritional factors in spinach.
Journal of environmental science and health. Part. B, Pesticides, food contaminants, and agricultural wastes (1983-08-01)
J Czapski, M Horbowicz, A Dobrzanski, J Bakowski
ABSTRACT
The influence of cycloate (active component of Ro-Neet 6E) on the biological value and residues in spinach was studied. During the growing season of spinach, cycloate residues decreased rapidly until by harvest time levels 0.0005--0.0008 mg/kg. had been reached. None of the studied herbicides had negative influence on the chemical composition of spinach.
Product Description
Supelco
Cycloate, PESTANAL®, analytical standard