MilliporeSigma
Cyclopeptoids: a novel class of phase-transfer catalysts.
Organic & biomolecular chemistry (2012-11-03)
Giorgio Della Sala, Brunello Nardone, Francesco De Riccardis, Irene Izzo
ABSTRACT
The synthesis, complexation properties and catalytic activities under phase-transfer (PT) conditions of differently substituted cyclohexapeptoids are reported. Association constants, for small cationic alkali, and catalytic performances, in a model nucleophilic substitution, are comparable to those of representative crown ethers. Noteworthy, the N-[2-(2-methoxyethoxy)ethyl] side chain derivative presents a catalytic efficiency comparable to that of crypt-222, and higher than some commonly used quaternary ammonium salts and crown ethers. Moreover its association constant for Na(+) complexation proved to be higher when compared with dicyclohexyl-18-crown-6. The synthesized cyclohexapeptoids represent the first example of these peptidomimetics in PT catalysis, anticipating interesting applications in biphasic PT methodology.
Sigma-Aldrich
Dicyclohexano-18-crown-6, 98%