MilliporeSigma
  • Furoquinoline alkaloids as photosensitizers in Chlamydomonas reinhardtii.
Furoquinoline alkaloids as photosensitizers in Chlamydomonas reinhardtii.
Mutation research (1991-07-01)
O Schimmer, I Kühne
ABSTRACT
Seven naturally occurring furoquinoline alkaloids were investigated for their photobiological activity using arg-1 cells of Chlamydomonas reinhardtii. UV-A-mediated toxicity of the compounds was calculated from the colony-forming ability of the treated cells. The UV-A-mediated mutagenicity was measured by counting the number of Arg+ revertants induced by the treatment. Dictamnine was found to be the strongest mutagen as well as the most toxic compound of the group. The mutagenic activities were measured as mutation frequencies at equal substance concentration and ranked in the following order: An increase in the number of substituents on the lateral aromatic nucleus greatly decreased the photomutagenicity. Except for evolitrine, a similar ranking order was found as reported for the dark mutagenicity of these compounds in Salmonella typhimurium strain TA98. Based on the result that furoquinolines are able to intercalate into DNA, we assume that the different mutagenic potencies may reflect differences in the geometry of the intercalation complex, which is important for the subsequent photochemical reaction.
Product Description
Supelco
Oil Orange SS, analytical standard