MilliporeSigma
  • O-Substituted derivatives of pralidoxime: muscarinic properties and protection against soman effects in rats.
O-Substituted derivatives of pralidoxime: muscarinic properties and protection against soman effects in rats.
European journal of pharmacology (2002-06-18)
Weng-Keong Loke, Meng-Kwoon Sim, Mei-Lin Go
ABSTRACT
O-Substituted aldoximes of the cholinesterase reactivator pralidoxime (O-methyl 1, O-benzyl 2, O-propynyl 3 and O-butynyl 4 derivatives) were synthesized and found to exhibit strong binding affinities for muscarinic receptors in rat brain, heart and submandibulary glands. The aldoximes were noncompetitive antagonists of acetylcholine-induced contraction of the guinea pig ileum. A good correlation was observed between binding affinity and pK(B). Weak anticholinesterase activities were observed for these compounds. When given intracerebroventricularly into conscious rats before soman administration (0.9 LD(50), subcutaneously), the aldoximes, like atropine but not pralidoxime, protected against respiratory depression (3,4) and bradycardia (2). No protection against soman-induced pressor effects was noted. The protective effects of these aldoximes may be the outcome of compensatory mechanisms, of which the cholinergic receptor agonist and antagonist properties of these compounds may be important.
Sigma-Aldrich
Acetaldehyde oxime, mixture of syn and anti, 99%