MilliporeSigma
  • Proteomic study of granulocytic differentiation induced by apigenin 7-glucoside in human promyelocytic leukemia HL-60 cells.
Proteomic study of granulocytic differentiation induced by apigenin 7-glucoside in human promyelocytic leukemia HL-60 cells.
European journal of nutrition (2011-11-25)
Eri Nakazaki, Soninkhishig Tsolmon, Junkyu Han, Hiroko Isoda
ABSTRACT
Nutritional factors is one of the most important regulators in the progression of cancer. Some dietary elements promote the growth of cancer but others, such as plant-derived compounds, may reverse this process. We tried to investigate yet another approach of cancer prevention through cancer cell differentiation, using a common non-mutagenic flavonoid apigenin 7-glucoside. HL-60 cells were treated with or without apigenin 7-glucoside. Cell proliferation was measured by MTT assay, and the cell cycle distribution was estimated by propidium iodide staining of DNA. To determine cellular differentiation, cell surface differentiation markers CD11b and CD14 were used. Two-dimensional gel electrophoresis was then performed to identify proteins that may be important in HL-60 cell differentiation following apigenin 7-glucoside treatment. Apigenin 7-glucoside inhibited HL-60 cell growth, dose- and time-dependently, but did not cause apoptosis. The distribution of cells at different stages in the cell cycle indicated an accumulation of treated cells in G(2)/M phase. Moreover, apigenin 7-glucoside induced granulocytic differentiation of HL-60 cells. Ten proteins that might play essential role in granulocytic differentiation were identified by proteomics. A complete understanding of the preventive effects of plant-based diet on cancer depends on the mechanisms of action of different plant components on processes. We hope these findings may contribute to the understandings of the different approaches for chemoprevention of cancer.
Product Description
Apigenin 7-glucoside, primary reference standard
Supelco
Apigenin 7-glucoside, analytical standard