MilliporeSigma
[Chemical constituents of Astragalus chinensis L].
Zhongguo Zhong yao za zhi = Zhongguo zhongyao zazhi = China journal of Chinese materia medica (1995-05-01)
Y Xu, L Wei
ABSTRACT
Three compounds have been isolated from the seeds of Astragalus chinensis. Their structures were identified on the basis of spectral data (UV, 13CNMR, 1HNMR, IR and MS) as octacosane, daucosterol and kaempferol.
Sigma-Aldrich
Octacosane, 99%