MilliporeSigma
  • Tubulin and benzimidazole-resistance in Trichostrongylus colubriformis (Nematoda).
Tubulin and benzimidazole-resistance in Trichostrongylus colubriformis (Nematoda).
The Journal of parasitology (1985-10-01)
N C Sangster, R K Prichard, E Lacey
ABSTRACT
Benzimidazole treatment produced greater effects on microtubule-dependent acetylcholinesterase secretion, the presence of microtubules in intestinal cells, and colchicine binding in susceptible compared with benzimidazole-resistant Trichostrongylus colubriformis. In addition, the binding of benzimidazoles was markedly reduced in preparations from the latter strain, indicating that the mechanism of resistance to benzimidazoles in this nematode involves a reduced affinity of tubulin for benzimidazoles.
Product Description
Supelco
Parbendazole, VETRANAL®, analytical standard