MilliporeSigma
Increase in life expectancy for mice fed diethylhydroxylamine (DEHA).
Journal of gerontology (1987-11-01)
J Heicklen, E Brown
ABSTRACT
ICR Swiss strain mice were fed diethylhydroxylamine (DEHA) for life starting at 44 days of age at 10, 30, 100, and 300 mg/kg-day. The 10 and 30 mg/kg-day males and the 10 mg/kg-day females showed an increase in mean age of death of 76 (12.7%), 45 (7.5%), and 51 (7.4%) days, respectively, compared to controls. The respective confidence levels are 99.2, 90.0, and 92.9% that these results are significant by Student's t test.
Sigma-Aldrich
N,N-Diethylhydroxylamine, ≥98%