MilliporeSigma
  • The metabolism of 1-methylhydantoin via 5-hydroxy-1-methylhydantoin in mammals.
The metabolism of 1-methylhydantoin via 5-hydroxy-1-methylhydantoin in mammals.
Biochimica et biophysica acta (1988-12-15)
K Ienaga, K Nakamura, F Naka, T Goto
ABSTRACT
The metabolic pathway of 1-methylhydantoin (2) via 5-hydroxy-1-methylhydantoin (3), methylparabanic acid (4) and N5-methyloxaluric acid (5) proved to be a major and general one in mammals. Hence the formation of (3), which has not been detected in normal tissue, is likely to be indirect in inflamed tissue, probably depending on the arising formation of (2) from creatinine (1).
Sigma-Aldrich
1-Methylhydantoin, 97%