MilliporeSigma
  • Activation of Ca2+-stimulated ATPase by phospholipid N-methylation in cardiac sarcoplasmic reticulum.
Activation of Ca2+-stimulated ATPase by phospholipid N-methylation in cardiac sarcoplasmic reticulum.
Biochemical and biophysical research communications (1985-07-16)
P K Ganguly, V Panagia, K Okumura, N S Dhalla
ABSTRACT
Incubation of cardiac sarcoplasmic reticulum (SR) in the presence of S-adenosyl-L-methionine, a methyl donor for the enzymatic N-methylation of phosphatidylethanolamine, increased Ca2+-stimulated ATPase activity. The increase in Ca2+-ATPase activity was not due to changes in the affinity for Ca2+ and was prevented by methyl acetimidate, an inhibitor of phospholipid N-methylation. The results suggest a possible regulatory role of phospholipid N-methylation in SR Ca2+-pump mechanism.
Sigma-Aldrich
Methyl acetimidate hydrochloride, technical grade