MilliporeSigma
  • M-cholinoreactivity of erythrocytes of non-pregnant and pregnant women evaluated by changes in the rate of erythrocyte agglutination under the influence of acetylcholine.
M-cholinoreactivity of erythrocytes of non-pregnant and pregnant women evaluated by changes in the rate of erythrocyte agglutination under the influence of acetylcholine.
Bulletin of experimental biology and medicine (2013-01-19)
A I Strelnikova, V I Tsirkin, A V Krysova, S V Hlybova, S L Dmitrieva
ABSTRACT
Acetylcholine (5.5×10(-10)-5.5×10(-6)M) accelerated erythrocyte agglutination in men, non-pregnant women in follicular phase of the menstrual cycle, and pregnant women in the first trimester. The effect was blocked with atropine (5.5×10(-6)M). Acetylcholine had no effect on the rate of erythrocyte agglutination in non-pregnant women in the luteal phase and pregnant women in the second and third trimesters, which coincided with the development of myometrium refractoriness to acetylcholine in pregnant women. The results indicate that erythrocytes can reflect M-cholinoreactivity of internal organs.
Product Description
Supelco
Atropine solution, 1.0 mg/mL in acetonitrile, ampule of 1 mL, certified reference material, Cerilliant®
Atropine for peak identification, European Pharmacopoeia (EP) Reference Standard