MilliporeSigma
  • Isolated central nervous system relapses in primary mediastinal large B-cell lymphoma after CHOP-like chemotherapy with or without Rituximab.
Isolated central nervous system relapses in primary mediastinal large B-cell lymphoma after CHOP-like chemotherapy with or without Rituximab.
Hematological oncology (2012-05-23)
Sotirios G Papageorgiou, Panayiotis Diamantopoulos, Georgia Levidou, Maria K Angelopoulou, Panagiota Economopoulou, Anna Efthimiou, Nikos Constantinou, Andreas Katsigiannis, Penelope Korkolopoulou, Vassiliki Pappa, Christina Economopoulou, George Georgiou, Maria Dimou, Panagiotis Tsirigotis, Marie-Christine Kyrtsonis, Ioannis Kotsianidis, Christina Kalpadakis, Meletios-Athanassios Dimopoulos, Photis Beris, John Meletis, Gerassimos A Pangalis, John Dervenoulas, Panayiotis Panayiotidis, Theodoros P Vassilakopoulos
ABSTRACT
Central nervous system (CNS) involvement in patients with primary mediastinal large B-cell (PMLBCL) lymphoma is a rare event, occurring in approximately 6% of patients, on the basis of the review of the literature prior to induction of Rituximab. The aim of this retrospective study was to describe the incidence of CNS relapse among 100 consecutive patients with PMLBCL who were treated with R-CHOP ± RT in comparison to patients treated with CHOP ± RT (n = 45) in 11 hospitals in Greece. Two patients experienced a CNS relapse, representing an overall frequency of 2.0% in R-CHOP treated patients and a 2-year actuarial incidence of 2.3%. Both patients had isolated CNS relapses. The incidence of CNS relapse after CHOP without Rituximab was 2/45 (4.4%) for a 2-year actuarial incidence of 7.5% (p = 0.29). Again, both patients had isolated CNS relapses. Parenchymal-only localizations accounted for 3/4 cases. Risk factors for CNS involvement could include leukocytosis, poor performance status and higher age-adjusted International Prognostic Index, although their impact was weakened by competing risk survival analysis. Both patients relapsing after R-CHOP required CNS radiotherapy to achieve a complete remission and be forwarded to high-dose therapy and autologous stem cell transplantation: They are both alive and disease-free 18 and 23 months after CNS relapse. Both cases relapsing after CHOP without Rituximab were salvaged by CNS radiotherapy (one also received intrathecal chemotherapy) entering long-term remissions. In conclusion, CNS relapses are rare in PMLBCL tending to be isolated in the CNS, probably reflecting the persistence of latent CNS disease than dissemination of resistant disease. The impact of Rituximab in reducing CNS relapses remains unknown. Established risk factors for CNS involvement in aggressive lymphomas may not be helpful in assessing the risk of CNS recurrence in this disease. Routine CNS prophylaxis is not probably required in PMLBCL.
Sigma-Aldrich
Thio-TEPA
Pricing and availability is not currently available.