MilliporeSigma
  • I2-mediated diversity oriented diastereoselective synthesis of amino acid derived trans-2,5-disubstituted morpholines, piperazines, and thiomorpholines.
I2-mediated diversity oriented diastereoselective synthesis of amino acid derived trans-2,5-disubstituted morpholines, piperazines, and thiomorpholines.
ACS combinatorial science (2011-12-01)
Saurav Bera, Gautam Panda
ABSTRACT
Diastereoselective trans-2,5-disubstituted amino acids derived diverse morpholines, piperazines and thiomorpholines were prepared in 30 min-1 h with high yields through iodine-mediated 6-exotrig type cyclization from a single common synthetic intermediate. The displacement of iodine with hydride ion gave a methyl substituent at the 2-position of morpholines which provides an additional opportunity for diversity oriented nucleophilic substitution on the rings as well as incorporation of substituents at the 5-position from amino acids constituents.
Sigma-Aldrich
Thiomorpholine, 98%