MilliporeSigma
  • Electrochemical oxide nanotube formation on the Ti-35Ta-xHf alloys for dental materials.
Electrochemical oxide nanotube formation on the Ti-35Ta-xHf alloys for dental materials.
Journal of nanoscience and nanotechnology (2011-11-23)
Byung-Hak Moon, Yong-Hoon Jeong, Han-Cheol Choe
ABSTRACT
In this study, we investigated the electrochemical oxide nanotube formation on the Ti-35Ta-xHf alloys for dental materials. The Ti-35Ta-xHf alloys contained from 3 wt.% to 15 wt.% Hf were manufactured by arc melting furnace. The nanotube oxide layers were formed on Ti-35Ta-xHf alloy by anodic oxidation method in 1 M H3PO4 electrolytes containing 0.5 wt.% NaF and 0.8 wt.% NaF at room temperature. The surface characteristics of Ti-35Ta-xHf alloy and nanotube morphology were determined by FE-SEM, STEM, and XRD. The nano-porous surface of Ti-35Ta-xHf alloys showed in 0.5 wt% NaF solution and nanotubular surface showed in 0.8 wt% NaF solution, respectively. The highly ordered nanotube layer without regular knots was formed on the Ti-35Ta-15Hf alloy in the 0.5 wt% NaF solution compared to on Ti-35Ta-3Hf and Ti-35Ta-7Hf alloys in 0.8 wt% NaF solution. Also, the nanotube length of Ti-35Ta-xHf alloys increased as Hf content increased.
Sigma-Aldrich
Hafnium, turnings, crystal bar, 99.7% trace metals basis