MilliporeSigma
  • Trimethylaurintricarboxylic acid inhibits human DNA methyltransferase 1: insights from enzymatic and molecular modeling studies.
Trimethylaurintricarboxylic acid inhibits human DNA methyltransferase 1: insights from enzymatic and molecular modeling studies.
Journal of molecular modeling (2011-08-02)
Jakyung Yoo, José L Medina-Franco
ABSTRACT
DNA methyltransferase 1 (DNMT1) is an emerging target for the treatment of cancer, brain disorders, and other diseases. Currently, there are only a few DNMT1 inhibitors with potential application as therapeutic agents or research tools. 5,5-Methylenedisalicylic acid is a novel scaffold previously identified by virtual screening with detectable although weak inhibitory activity of DNMT1 in biochemical assays. Herein, we report enzyme inhibition of a structurally related compound, trimethylaurintricarboxylic acid (NSC97317) that showed a low micromolar inhibition of DNMT1 (IC(50) = 4.79 μM). Docking studies of the new inhibitor with the catalytic domain of DNMT1 suggest that NSC97317 can bind into the catalytic site. Interactions with amino acid residues that participate in the mechanism of DNA methylation contribute to the binding recognition. In addition, NSC97317 had a good match with a structure-based pharmacophore model recently developed for inhibitors of DNMT1. Trimethylaurintricarboxylic acid can be a valuable biochemical tool to study DNMT1 inhibition in cancer and other diseases related to DNA methylation.
Sigma-Aldrich
Aluminon, JIS special grade
Sigma-Aldrich
Aurintricarboxylic acid, practical grade, ≥85% (titration), powder
Sigma-Aldrich
Aurintricarboxylic acid ammonium salt, ACS reagent
Sigma-Aldrich
Aurintricarboxylic acid ammonium salt, powder