MilliporeSigma
  • Simple synthesis of 1,3-cyclopentanedione derived probes for labeling sulfenic acid proteins.
Simple synthesis of 1,3-cyclopentanedione derived probes for labeling sulfenic acid proteins.
Chemical communications (Cambridge, England) (2011-07-09)
Jiang Qian, Chananat Klomsiri, Marcus W Wright, S Bruce King, Allen W Tsang, Leslie B Poole, Cristina M Furdui
ABSTRACT
Facile, two-step synthesis and kinetic characterization of new chemical probes for selective labeling of sulfenic acid (-SOH) in proteins are presented. The synthesis route relies on the simple and highly efficient Michael addition of thiol containing tags or linkers to 4-cyclopentene-1,3-dione, the unsaturated derivative of 1,3-cyclopentanedione.
Sigma-Aldrich
1,3-Cyclopentanedione, 97%
Sigma-Aldrich
4-Cyclopentene-1,3-dione, 95%