MilliporeSigma
On the interaction between synthetic detergents and enzymatic proteins.
Canadian journal of biochemistry and cell biology = Revue canadienne de biochimie et biologie cellulaire (1984-01-01)
C Treves, M T Vincenzini, F Favilli, P Vanni, V Baccari
ABSTRACT
Eight highly purified and crystalline enzymes representative of fundamental metabolic pathways and of cellular structure were tested with seven detergents. The enzyme-detergent interaction was studied as a time-independent (inhibition) or time-dependent (inactivation) phenomenon. Our results imply a specificity of detergent action on the enzymes studied.
Product Description
Supelco
Sodium 1-octanesulfonate monohydrate, ≥99.0% (T)