MilliporeSigma
  • [Studies on the chemical constituents of the petroleum ether portion of Nervilia fordii].
[Studies on the chemical constituents of the petroleum ether portion of Nervilia fordii].
Zhong yao cai = Zhongyaocai = Journal of Chinese medicinal materials (2010-09-30)
Han-shen Zhen, Qin Qiu, Huan-heng Mo, Ye-fei Yuan, Yan-yuan Zhou, Min-qi Huang
ABSTRACT
To study the chemical constituents of the petroleum ether portion of Nervilia fordii. The constituents were separated and purified by using column chromatography with silica gel. These compounds were identified by their physical and spectral data. Six compounds were isolated and identified as Cyclohomonervilol (I), Octacosanoicacid (II), Stigmasterol (III), Cyclohomonervilol-(E)-p-hydroxy cinnamate (IV), 24(R/alpha)-dihydrocycloeucalenol-(E)-p-hydroxy cinnamate (V), Docosanoic acid (VI). Compounds I-VI are isolated from this plant for the first time.
Product Description
Supelco
Behenic acid, analytical standard
Sigma-Aldrich
Behenic acid, 99%
Sigma-Aldrich
Octacosanoic acid, synthetic, ≥98%