MilliporeSigma
  • Mutation of charged residues to neutral ones accelerates urea denaturation of HP-35.
Mutation of charged residues to neutral ones accelerates urea denaturation of HP-35.
The journal of physical chemistry. B (2010-08-24)
Haiyan Wei, Lijiang Yang, Yi Qin Gao
ABSTRACT
Following the studies of urea denaturation of β-hairpins using molecular dynamics, in this paper, molecular dynamics simulations of two peptides, a 35 residue three helix bundle villin headpiece protein HP-35 and its doubly norleucine-substituent mutant (Lys24Nle/Lys29Nle) HP-35 NleNle, were undertaken in urea solutions to understand the molecular mechanism of urea denaturation of α-helices. The mutant HP-35 NleNle was found to denature more easily than the wild type. During the expansion of the small hydrophobic core, water penetration occurs first, followed by that of urea molecules. It was also found that the initial hydration of the peptide backbone is achieved through water hydrogen bonding with the backbone CO groups during the denaturation of both polypeptides. The mutation of the two charged lysine residues to apolar norleucine enhances the accumulation of urea near the hydrophobic core and facilitates the denaturation process. Urea also interacts directly with the peptide backbone as well as side chains, thereby stabilizing nonnative conformations. The mechanism revealed here is consistent with the previous study on secondary structure of β-hairpin polypeptide, GB1, PEPTIDE 1, and TRPZIP4, suggesting that there is a general mechanism in the denaturation of protein backbone hydrogen bonds by urea.
Sigma-Aldrich
L-Norleucine, suitable for amino acid analysis, BioReagent
Sigma-Aldrich
L-Norleucine, ≥98% (TLC)