MilliporeSigma
  • Metabolism and absorption of auraptene (7-geranyloxylcoumarin) in male SD rats: comparison with 7-ethoxycoumarin.
Metabolism and absorption of auraptene (7-geranyloxylcoumarin) in male SD rats: comparison with 7-ethoxycoumarin.
Nutrition and cancer (2008-04-30)
Wataru Kuki, Keisuke Hosotani, Hajime Ohigashi, Akira Murakami
ABSTRACT
We previously reported that auraptene (7-geranyloxycoumarin; AUR), a coumarin that occurs widely in citrus fruit, has been shown to be a promising cancer-preventive agent in several rodent models. However, its bioavailability and metabolism have not been investigated. In this study, we compared the metabolism characteristics of AUR with those of 7-ethoxycoumarin (ETC) in male Sprague Dawley rats. Each (500 micromol/kg body weight) was given separately by a single gastric intubation procedure, and digestive tract, liver, and kidney were removed at 1, 4, and 24 h after administration. The localization profiles of AUR and ETC in the gastrointestinal tract were similar. However, AUR, in contrast to ETC, showed significant localization in the liver from 1 to 4 h. Treatments of serum and urinary samples with glucuronidase/sulfatase led to the detection of significant amounts of umbelliferone (7-hydroxycoumarin; UMB), and serum and urinary concentrations of UMB following ETC administration were significantly higher than with AUR administration. Our results suggest that AUR, which bears a geranyloxyl side chain, has a longer life span than ETC, and this property may be associated with its previously reported chemopreventive and xenobiotics metabolizing activities.
Sigma-Aldrich
7-Ethoxycoumarin