MilliporeSigma
  • Evaluation of the genotoxic and antioxidant effects of two novel feed additives (ethoxyquin complexes with flavonoids) by the comet assay and micronucleus test.
Evaluation of the genotoxic and antioxidant effects of two novel feed additives (ethoxyquin complexes with flavonoids) by the comet assay and micronucleus test.
Food additives and contaminants (2007-05-10)
A Błaszczyk, J Skolimowski
ABSTRACT
The complexes of antioxidant ethoxyquin (1,2-dihydro-6-ethoxy-2,2,4-trimethylquinoline; EQ) with rutin or quercetin (EQ-R and EQ-Q, respectively) were studied in human lymphocytes for genotoxic and antioxidant activities with the use of the comet assay and micronucleus test. The study was undertaken to search for new potential antioxidants, and was motivated by reports of unfavourable side-effects observed in animals fed with feeds containing EQ, which is allowed up to 150 mg kg(-1) (0.015%) in complete animal feed. It was shown that EQ-R induced DNA damage in human lymphocytes when used at all the concentrations studied (1-25 microM), while after EQ-Q treatment, the genotoxic effect was observed mainly after higher doses (10 and 25 microM). An increase in the number of micronuclei was observed only for EQ-Q after a dose of 50 microM. The studied compounds decreased the degree of DNA damage induced by hydrogen peroxide (10 microM) in the comet assay. The results obtained in both tests showed that the antioxidant activity of EQ-Q was comparable with that of EQ, so further detailed studies are necessary to estimate its possible usefulness as a feed preservative.
Product Description
Supelco
Ethoxyquin, PESTANAL®, analytical standard
Sigma-Aldrich
Ethoxyquin, ≥75% (capillary GC)