MilliporeSigma
  • Synthesis, tubulin binding, antineoplastic evaluation, and structure-activity relationship of oncodazole analogues.
Synthesis, tubulin binding, antineoplastic evaluation, and structure-activity relationship of oncodazole analogues.
Journal of medicinal chemistry (1989-02-01)
L I Kruse, D L Ladd, P B Harrsch, F L McCabe, S M Mong, L Faucette, R Johnson
ABSTRACT
In an attempt to identify a soluble oncodazole analogue that could be easily formulated, a series of substituted oncodazoles was synthesized and evaluated for tubulin binding affinity, in vitro cytotoxicity against cultured mouse B-16 cells, and ability to prolong lifespan at the maximally tolerated dose in the P388 mouse leukemia model. Biological evaluation of all the isomeric methyloncodazoles demonstrated the thiophene 4'-position to be the only site of significant bulk tolerance, although substitution of this position with polar or charged functional groups abolished biological activity. Simple esters of the 4'-carboxymethyloncodazole were shown to have enhanced antitumor activity and tubulin binding affinity relative to oncodazole. Despite a failure of this study to identify a water-soluble oncodazole with antitumor activity, the structure-activity relationship developed led to a derivative with enhanced activity in the P388 leukemia model and facilitated the preparation of a biologically active photolabile analogue.
Sigma-Aldrich
N-Boc-1,4-butanediamine, ≥97.0% (GC/NT)
Pricing and availability is not currently available.